4-Carbamoyltetrahydropyran 98%
Supplier: Apollo Scientific
|
Warning
|
OR9367-1G
OR9367-5G
APOSOR9367-1GEA
55.1
EUR
APOSOR9367-1G
APOSOR9367-5G
4-Carbamoyltetrahydropyran 98%
4-Carbamoyltetrahydropyran
Formula:
C₆H₁₁NO₂ Melting Pt: 181…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD08235066 CAS Number: 344329-76-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...