Ethyl-4,5,6,7-tetrahydroisoindole-1-carboxylate 95%

Supplier: Apollo Scientific

OR937721-1G OR937721-250MG
APOSOR937721-1GEA 230 EUR
APOSOR937721-1G APOSOR937721-250MG
Ethyl-4,5,6,7-tetrahydroisoindole-1-carboxylate 95%
Ethyl-4,5,6,7-tetrahydroisoindole-1-carboxylate
Formula: C₁₁H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD06227773
CAS Number: 65880-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR