3-Pyridazineacetonitrile

Supplier: Apollo Scientific

OR946599-1G OR946599-250MG OR946599-500MG
APOSOR946599-1GEA 1800 EUR
APOSOR946599-1G APOSOR946599-250MG APOSOR946599-500MG
3-Pyridazineacetonitrile
3-Pyridazineacetonitrile
Formula: C₆H₅N₃
Storage Temperature: Ambient
MDL Number: MFCD17015868
CAS Number: 27349-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR