3'-Ethynylacetanilide 95%
Supplier: Apollo Scientific
|
Warning
|
OR9855-1G
OR9855-5G
OR9855-250MG
APOSOR9855-1GEA
103
EUR
APOSOR9855-1G
APOSOR9855-5G
APOSOR9855-250MG
3'-Ethynylacetanilide 95%
3'-Ethynylacetanilide
Formula:
C₁₀H₉NO MW: 159.1846 g/mol Melting Pt: 92…93 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00831583 CAS Number: 70933-58-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...