4-Bromo-α,α,α,2,3,5,6-heptafluorotoluene 98%

Warning

PC1560E-100G PC1560E-25G
APOSPC1560E-100GEA 764 EUR
APOSPC1560E-100G APOSPC1560E-25G
4-Bromo-α,α,α,2,3,5,6-heptafluorotoluene 98%
4-Bromo-α,α,α,2,3,5,6-heptafluorotoluene
Formula: C₇BrF₇
Boiling Pt: 152 °C
Density: 1,94 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00075240
CAS Number: 17823-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR