2-Bromo-5-fluoropyrazine 98%

Supplier: Apollo Scientific

PC201057-100MG PC201057-1G PC201057-250MG
APOSPC201057-100MGEA 42.2 EUR
APOSPC201057-100MG APOSPC201057-1G APOSPC201057-250MG
2-Bromo-5-fluoropyrazine 98%
2-Bromo-5-fluoropyrazine
Formula: C₄H₂BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD14702726
CAS Number: 1209459-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR