3-Aminomethyl-3-fluorooxetane

Supplier: Apollo Scientific
Warning

PC406004-100MG PC406004-250MG
APOSPC406004-100MGEA 152 EUR
APOSPC406004-100MG APOSPC406004-250MG
3-Aminomethyl-3-fluorooxetane
3-Aminomethyl-3-fluorooxetane
Formula: C₄H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD18073259
CAS Number: 883311-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR