4-n-Butylbenzotrifluoride 95%
Supplier: Apollo Scientific
PC520709-10G
PC520709-5G
APOSPC520709-10GEA
1120
EUR
APOSPC520709-10G
APOSPC520709-5G
4-n-Butylbenzotrifluoride 95%
4-n-Butylbenzotrifluoride
Formula:
C₁₁H₁₃F₃ Storage Temperature: Ambient |
MDL Number:
MFCD14525528 CAS Number: 61342-04-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...