Perfluoroacetophenone
Supplier: Apollo Scientific
|
Warning
|
PC7644-5G
PC7644-1G
APOSPC7644-5GEA
190
EUR
APOSPC7644-5G
APOSPC7644-1G
Perfluoroacetophenone
Perfluoroacetophenone
Formula:
C₈F₈O Boiling Pt: 130…131 °C Density: 1,609 g/cm³ (25 °C) Flash Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216596 CAS Number: 652-22-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...