Fluorotriphenylsilane
Supplier: Apollo Scientific
|
Danger
|
PC7870-5G
PC7870-25G
APOSPC7870-5GEA
78.3
EUR
APOSPC7870-5G
APOSPC7870-25G
Fluorotriphenylsilane
Fluorotriphenylsilane
Formula:
C₁₈H₁₅FSi MW: 278.4 g/mol Boiling Pt: 210 °C/10mmHg Melting Pt: 64 °C Density: 1.212 Storage Temperature: Ambient |
MDL Number:
MFCD00017899 CAS Number: 379-50-0 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...