Lloyd and McCown's woody plant medium with vitamins
Supplier: Apollo Scientific
PML449B-10L
PML449C-50L
APOSPML449B-10LEA
68
EUR
APOSPML449B-10L
APOSPML449C-50L
Lloyd and McCown's woody plant medium with vitamins
Cell Culture Media
Contains the WPM macro- and micronutrients and vitamins as described by Lloyd and McCown (1981). Plant tissue culture tested.