Display unit to monitor connected fill-level sensors in b.safe Waste Caps and b.safe Waste Caps.
- With plugs for EU, Australia, USA, Japan and UK
- Compatible with any version of b.safe Waste Caps
- Additional comfort provides the possibility to snooze the alarm tone during the canister exchange
Alarm box with aluminium case for installation in the front plates of lab furniture. Front side with touch buttons and LED indicators to monitor the operation and the fill level of the connected sensors. Optical and acoustic main alarm when containers are full. The acoustic main alarm can be deactivated for 3 minutes until an empty container is confirmed. Suitable for b.safe Waste Caps LC / System Waste Caps LC with main alarm and b.safe Waste Caps DLC / System Waste Caps DLC with pre and main alarm. Includes power supply (input: AC 100 to 240 V/50 to 60 Hz, 0,5 A, output: 24 V DC, 0,5 A) with plugs for EU, Australia, USA, Japan, and UK.
Aluminium casing allows installation in the front plates of lab furniture. Easy operation via touch surface.
Accessories information: Combined Signal Device with red LED flashlight and additional siren (continuous tone) can be used in a separate room or at high noise levels. Depending on the connected output socket at the b.safe Display it can be used as single alarm or to monitor several containers. IP 65-protected housing made of ABS. Includes power supply (input: AC 100 to 240 V/50 to 60 Hz, 5,5 A, output: 24 V DC, 0,5 A) and plugs for EU, Australia, USA, Japan, and UK.