L-Erythrulose

Supplier: ACROS ORGANICS

467300250 467301000
ACRO467300250EA 119 EUR
ACRO467300250 ACRO467301000
L-Erythrulose
L-Erythrulose
Formula: C₄H₈O₄
MW: 120.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004703
CAS Number: 533-50-6

Order Now

Specification Test Results

Appearance Yellow to green to brown viscous liquid
Infrared spectrum Conforms
HPLC ≥ 76.0%
Water ≤ 24.0%
Sulfated ash ≤ 4.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR