1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone 98%

BD0055-10G BD0055-1G
BLDPBD0055-10GEA 58.8 EUR
BLDPBD0055-10G BLDPBD0055-1G
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone 98%
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)ethanone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00006823
CAS Number: 2879-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR