6-Bromo-4-(trifluoromethyl)nicotinonitrile 97%

BD00673680-100 BD00673680-250
BLDPBD00673680-100EA 215 EUR
BLDPBD00673680-100 BLDPBD00673680-250
6-Bromo-4-(trifluoromethyl)nicotinonitrile 97%
6-Bromo-4-(trifluoromethyl)nicotinonitrile

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₂BrF₃N₂
Storage Temperature: Refrigerator
MDL Number: MFCD28737575
CAS Number: 1807025-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR