Boc-L-octahydroindole-2-carboxylic acid 97%

BD00745759-25G BD00745759-1G
BLDPBD00745759-25GEA 335 EUR
BLDPBD00745759-25G BLDPBD00745759-1G
Boc-L-octahydroindole-2-carboxylic acid 97%
Boc-L-octahydroindole-2-carboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₄H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD09750486
CAS Number: 109523-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR