Tris(dibenzylideneacetone)dipalladium(0) 98%

BD00768014-1G BD00768014-5G
BLDPBD00768014-1GEA 97.7 EUR
BLDPBD00768014-1G BLDPBD00768014-5G
Tris(dibenzylideneacetone)dipalladium(0) 98%
Tris(dibenzylideneacetone)dipalladium(0)

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₅₁H₄₂O₃Pd₂
Storage Temperature: Ambient
MDL Number: MFCD00013310
CAS Number: 52409-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR