8-Bromo-4-chloro-5-methylquinazoline 95%

BD00779556-100 BD00779556-1G
BLDPBD00779556-100EA 224 EUR
BLDPBD00779556-100 BLDPBD00779556-1G
8-Bromo-4-chloro-5-methylquinazoline 95%
8-Bromo-4-chloro-5-methylquinazoline

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₆BrClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD31699367
CAS Number: 1566908-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR