3,3'-Disulphanediylbis(propan-1-ol) 97%

BD00798528-250 BD00798528-100
BLDPBD00798528-250EA 71.4 EUR
BLDPBD00798528-250 BLDPBD00798528-100
3,3'-Disulphanediylbis(propan-1-ol) 97%
3,3'-Disulphanediylbis(propan-1-ol)

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₁₄O₂S₂
Storage Temperature: Ambient
MDL Number: MFCD01630941
CAS Number: 30453-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR