2-(3,4-Dichloro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
|
|
BD00799699-1G
BD00799699-100
BLDPBD00799699-1GEA
185
EUR
BLDPBD00799699-1G
BLDPBD00799699-100
2-(3,4-Dichloro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(3,4-Dichloro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₃H₁₇BCl₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD22415026 CAS Number: 2121513-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...