VU 0365114 98%

BD00802581-10M BD00802581-50M
BLDPBD00802581-10MEA 335 EUR
BLDPBD00802581-10M BLDPBD00802581-50M
VU 0365114 98%
VU 0365114

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂₂H₁₄F₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD22683818
CAS Number: 1208222-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR