(R)-1,3-Dimethylpiperazin-2-one 98%
|
|
BD00803217-250
BD00803217-100
BLDPBD00803217-250EA
347
EUR
BLDPBD00803217-250
BLDPBD00803217-100
(R)-1,3-Dimethylpiperazin-2-one 98%
(R)-1,3-Dimethylpiperazin-2-one
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₆H₁₂N₂O Storage Temperature: Ambient |
MDL Number:
MFCD22201139 CAS Number: 1240300-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...