5-Ethynylpyridine-3-carboxylic acid 95%
|
|
BD00829416-1G
BD00829416-250
BLDPBD00829416-1GEA
159
EUR
BLDPBD00829416-1G
BLDPBD00829416-250
5-Ethynylpyridine-3-carboxylic acid 95%
5-Ethynylpyridine-3-carboxylic acid
Storage condition: Sealed in dry, 2-8°C
Formula:
C₈H₅NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD18207405 CAS Number: 1211533-87-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...