(R)-1-(5-Bromo-4-methylpyridin-2-yl)ethan-1-amine dihydrochloride 98+%
|
|
BD00834112-100
BD00834112-250
BLDPBD00834112-100EA
320
EUR
BLDPBD00834112-100
BLDPBD00834112-250
(R)-1-(5-Bromo-4-methylpyridin-2-yl)ethan-1-amine dihydrochloride 98+%
(R)-1-(5-Bromo-4-methylpyridin-2-yl)ethan-1-amine dihydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₈H₁₃BrCl₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD31758480 CAS Number: 2703664-73-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...