(5-Fluoro-2-nitrophenyl)methanamine hcl 95%
|
|
BD00862502-250
BD00862502-1G
BLDPBD00862502-250EA
397
EUR
BLDPBD00862502-250
BLDPBD00862502-1G
(5-Fluoro-2-nitrophenyl)methanamine hcl 95%
(5-Fluoro-2-nitrophenyl)methanamine hcl
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₇H₈ClFN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD13194381 CAS Number: 1214366-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...