3-(Oxan-4-yl)phenol 95%
|
|
BD00873806-100
BD00873806-1G
BLDPBD00873806-100EA
244
EUR
BLDPBD00873806-100
BLDPBD00873806-1G
3-(Oxan-4-yl)phenol 95%
3-(Oxan-4-yl)phenol
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₁H₁₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD21099827 CAS Number: 1353854-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...