2-(tert-Butoxycarbonylamino)pyridin-4-ylboronic acid 97%

BD00928676-1G BD00928676-250
BLDPBD00928676-1GEA 233 EUR
BLDPBD00928676-1G BLDPBD00928676-250
2-(tert-Butoxycarbonylamino)pyridin-4-ylboronic acid 97%
2-(tert-Butoxycarbonylamino)pyridin-4-ylboronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₀H₁₅BN₂O₄
Storage Temperature: Freezer
MDL Number: MFCD13188969
CAS Number: 903513-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR