Ethyl 2-methyl-3-oxopentanoate 95%

BD00945661-100 BD00945661-250
BLDPBD00945661-100EA 205 EUR
BLDPBD00945661-100 BLDPBD00945661-250
Ethyl 2-methyl-3-oxopentanoate 95%
Ethyl 2-methyl-3-oxopentanoate

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₁₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00032120
CAS Number: 759-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR