Adamantane-1,3-diyldimethanamine dihydrochloride 97%

BD00950412-250 BD00950412-25G
BLDPBD00950412-250EA 34.7 EUR
BLDPBD00950412-250 BLDPBD00950412-25G
Adamantane-1,3-diyldimethanamine dihydrochloride 97%
Adamantane-1,3-diyldimethanamine dihydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₂H₂₄Cl₂N₂
Storage Temperature: Ambient
CAS Number: 69887-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR