Ethyl 4-hydroxy-5-(trifluoromethyl)quinoline-3-carboxylate 98%

BD00974884-100 BD00974884-1G
BLDPBD00974884-100EA 191 EUR
BLDPBD00974884-100 BLDPBD00974884-1G
Ethyl 4-hydroxy-5-(trifluoromethyl)quinoline-3-carboxylate 98%
Ethyl 4-hydroxy-5-(trifluoromethyl)quinoline-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₃H₁₀F₃NO₃
MW: 285.2186 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28678830
CAS Number: 56881-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR