1-Cyclopropylethane-1,2-diol 95%
|
|
BD01023395-250
BD01023395-100
BLDPBD01023395-250EA
237
EUR
BLDPBD01023395-250
BLDPBD01023395-100
1-Cyclopropylethane-1,2-diol 95%
1-Cyclopropylethane-1,2-diol
Storage condition: Sealed in dry, 2-8°C
Formula:
C₅H₁₀O₂ MW: 102.1317 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD24230899 CAS Number: 784105-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...