Methyl 5-(fluorosulphonyl)-2-hydroxybenzoate 95%

BD01024674-250 BD01024674-1G
BLDPBD01024674-250EA 92.4 EUR
BLDPBD01024674-250 BLDPBD01024674-1G
Methyl 5-(fluorosulphonyl)-2-hydroxybenzoate 95%
Methyl 5-(fluorosulphonyl)-2-hydroxybenzoate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₇FO₅S
Storage Temperature: Refrigerator
MDL Number: MFCD28968600
CAS Number: 400-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR