3-Amino-1-phenylbutan-2-one hydrochloride 95%
|
|
BD01042965-250
BD01042965-100
BLDPBD01042965-250EA
192
EUR
BLDPBD01042965-250
BLDPBD01042965-100
3-Amino-1-phenylbutan-2-one hydrochloride 95%
3-Amino-1-phenylbutan-2-one hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₀H₁₄ClNO Storage Temperature: Ambient |
MDL Number:
MFCD20134012 CAS Number: 191104-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...