(S)-2-(Piperazin-2-yl)acetonitrile dihydrochloride 97%

BD01092913-5G BD01092913-250
BLDPBD01092913-5GEA 443 EUR
BLDPBD01092913-5G BLDPBD01092913-250
(S)-2-(Piperazin-2-yl)acetonitrile dihydrochloride 97%
(S)-2-(Piperazin-2-yl)acetonitrile dihydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₆H₁₃Cl₂N₃
Storage Temperature: Refrigerator
MDL Number: MFCD32173174
CAS Number: 2158301-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR