5,5'-(Pyridine-3,5-diyl)diisophthalic acid 98%

BD01099811-100 BD01099811-250
BLDPBD01099811-100EA 64.1 EUR
BLDPBD01099811-100 BLDPBD01099811-250
5,5'-(Pyridine-3,5-diyl)diisophthalic acid 98%
5,5'-(Pyridine-3,5-diyl)diisophthalic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂₁H₁₃NO₈
Storage Temperature: Ambient
CAS Number: 1433029-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR