3-Amino-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-chromen-2-one 97%
|
|
BD01104706-250
BD01104706-100
BLDPBD01104706-250EA
486
EUR
BLDPBD01104706-250
BLDPBD01104706-100
3-Amino-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-chromen-2-one 97%
3-Amino-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-chromen-2-one
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formula:
C₁₅H₁₈BNO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD32644527 CAS Number: 2828431-91-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...