1,2-Dimethyl-3-octyl-1H-imidazol-3-ium bromide 98%

BD01107731-5G BD01107731-100
BLDPBD01107731-5GEA 13.7 EUR
BLDPBD01107731-5G BLDPBD01107731-100
1,2-Dimethyl-3-octyl-1H-imidazol-3-ium bromide 98%
1,2-Dimethyl-3-octyl-1H-imidazol-3-ium bromide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₃H₂₅BrN₂
Storage Temperature: Ambient
MDL Number: MFCD19440741
CAS Number: 61546-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR