5-(Trifluoromethyl)-2,3-dihydro-1H-inden-1-amine hydrochloride 95+%

BD01131589-100 BD01131589-250
BLDPBD01131589-100EA 355 EUR
BLDPBD01131589-100 BLDPBD01131589-250
5-(Trifluoromethyl)-2,3-dihydro-1H-inden-1-amine hydrochloride 95+%
5-(Trifluoromethyl)-2,3-dihydro-1H-inden-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₀H₁₁ClF₃N
Storage Temperature: Ambient
MDL Number: MFCD30725597
CAS Number: 936220-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR