(S)-1-(3-(Trifluoromethoxy)phenyl)ethanamine hydrochloride 95+%

BD01141563-250 BD01141563-1G
BLDPBD01141563-250EA 334 EUR
BLDPBD01141563-250 BLDPBD01141563-1G
(S)-1-(3-(Trifluoromethoxy)phenyl)ethanamine hydrochloride 95+%
(S)-1-(3-(Trifluoromethoxy)phenyl)ethanamine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₉H₁₁ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD12757220
CAS Number: 1391567-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR