Thianthrene 5-oxide 97%

BD01143199-100 BD01143199-1G
BLDPBD01143199-100EA 57.8 EUR
BLDPBD01143199-100 BLDPBD01143199-1G
Thianthrene 5-oxide 97%
Thianthrene 5-oxide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₂H₈OS₂
Storage Temperature: Ambient
MDL Number: MFCD01464038
CAS Number: 2362-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR