1-(Adamantan-1-yl)-3-mesityl-4,5-dihydro-1H-imidazol-3-ium chloride 97%

BD01143339-50M BD01143339-1G
BLDPBD01143339-50MEA 43.1 EUR
BLDPBD01143339-50M BLDPBD01143339-1G
1-(Adamantan-1-yl)-3-mesityl-4,5-dihydro-1H-imidazol-3-ium chloride 97%
1-(Adamantan-1-yl)-3-mesityl-4,5-dihydro-1H-imidazol-3-ium chloride

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₂₂H₃₁ClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD08276859
CAS Number: 639820-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR