3-Bromo-2,5,6-trifluorobenzonitrile 97%

BD01160959-250 BD01160959-100
BLDPBD01160959-250EA 101 EUR
BLDPBD01160959-250 BLDPBD01160959-100
3-Bromo-2,5,6-trifluorobenzonitrile 97%
3-Bromo-2,5,6-trifluorobenzonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇HBrF₃N
Storage Temperature: Ambient
MDL Number: MFCD30830098
CAS Number: 485318-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR