4-Cyanophenyl trifluoromethanesulphonate 98%
|
|
BD01185283-25G
BD01185283-100
BLDPBD01185283-25GEA
998
EUR
BLDPBD01185283-25G
BLDPBD01185283-100
4-Cyanophenyl trifluoromethanesulphonate 98%
4-Cyanophenyl trifluoromethanesulphonate
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₈H₄F₃NO₃S MW: 251.1825 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00973310 CAS Number: 66107-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...