Dibromo(1,10-phenanthroline-κN1,κN10)nickel 98%

BD01224038-5G BD01224038-25G
BLDPBD01224038-5GEA 118 EUR
BLDPBD01224038-5G BLDPBD01224038-25G
Dibromo(1,10-phenanthroline-κN1,κN10)nickel 98%
Dibromo(1,10-phenanthroline-κN1,κN10)nickel

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₂H₈Br₂N₂Ni
Storage Temperature: Refrigerator
CAS Number: 48165-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR