4-Cyclohexyl-2,6-dimethoxyphenol 97%

BD01285797-250 BD01285797-100
BLDPBD01285797-250EA 706 EUR
BLDPBD01285797-250 BLDPBD01285797-100
4-Cyclohexyl-2,6-dimethoxyphenol 97%
4-Cyclohexyl-2,6-dimethoxyphenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₄H₂₀O₃
Storage Temperature: Ambient
MDL Number: MFCD33022389
CAS Number: 2682114-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR