Dimethyl 4,5-dimethylphthalate 98%
|
|
BD01290210-250
BD01290210-5G
BLDPBD01290210-250EA
52.5
EUR
BLDPBD01290210-250
BLDPBD01290210-5G
Dimethyl 4,5-dimethylphthalate 98%
Dimethyl 4,5-dimethylphthalate
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₂H₁₄O₄ MW: 222.2372 g/mol Storage Temperature: Ambient |
CAS Number:
17649-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...