3-Methylquinoline-7-carboxylic acid hydrochloride 97%

BD01306808-100 BD01306808-250
BLDPBD01306808-100EA 308 EUR
BLDPBD01306808-100 BLDPBD01306808-250
3-Methylquinoline-7-carboxylic acid hydrochloride 97%
3-Methylquinoline-7-carboxylic acid hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₁H₁₀ClNO₂
Storage Temperature: Ambient
CAS Number: 2757731-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR