Benzothiazole-5-carboxylic acid 95%
|
|
BD01314454-250
BD01314454-25G
BLDPBD01314454-250EA
30.5
EUR
BLDPBD01314454-250
BLDPBD01314454-25G
Benzothiazole-5-carboxylic acid 95%
Benzothiazole-5-carboxylic acid
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₅NO₂S MW: 179.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06796380 CAS Number: 68867-17-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...