3-Hydroxypropyl 3,4-difluorobenzoate 98%

BD01339254-250 BD01339254-5G
BLDPBD01339254-250EA 11.6 EUR
BLDPBD01339254-250 BLDPBD01339254-5G
3-Hydroxypropyl 3,4-difluorobenzoate 98%
3-Hydroxypropyl 3,4-difluorobenzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
CAS Number: 684648-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR