N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-O-allyl-L-threonine 98%

BD01383143-250 BD01383143-100
BLDPBD01383143-250EA 154 EUR
BLDPBD01383143-250 BLDPBD01383143-100
N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-O-allyl-L-threonine 98%
N-(((9H-Fluoren-9-yl)methoxy)carbonyl)-O-allyl-L-threonine
Formula: C₂₂H₂₃NO₅
Storage Temperature: Refrigerator
CAS Number: 1945973-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR